Learn More
ATCC Leishmania mexicana (Biagi) Garnham, GFP-expressing promastigote strain MNYC/BZ/62/M379
Supplier: ATCC PRA416
Leishmania mexicana (Biagi) Garnham is a GFP-expressing promastigote strain (strain designation MNYC/BZ/62/M379) supplied as cryopreserved cells for infectious disease research and assay development. Maintain axenic cultures at 25°C under selective conditions and follow recommended thawing and cryopreservation procedures.
- GFP-expressing promastigotes suitable for visualization and assay work.
- Axenic culture; incubate at 25°C.
- Medium: M199, modified, with 300 μg/mL G418 for selection.
- Storage: -80°C for short-term, vapor phase of liquid nitrogen for long-term.
- Propagation: transfer every 7-14 days; follow recommended thaw and cryopreservation steps.
- Isolation: animal source (Sumichrast's vesper rat), Belize, 1962.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.