Learn More
ATCC Leishmania tarentolae Parrot (strain Tar II), frozen axenic culture for research (ATCC 30267)
Supplier:  ATCC 30267
Encompass
Leishmania tarentolae Parrot (strain Tar II) is a saurian-associated parasitic protozoan provided as a frozen, cryopreserved culture for laboratory research. The strain is maintained axenically and is suited to molecular biology, parasitology, and recombinant protein expression workflows.
  • Axenic parasitic protozoan strain isolated from Tarentola mauritanica (lizard).
  • Supplied cryopreserved for long-term storage and recovery to culture.
  • Cultured in diphasic blood agar medium; incubate at 25°C with the cap secured.
  • Biosafety level 1 handling requirements.
  • Suitable as a nonpathogenic model for Leishmania biology and protein expression studies.
Catalog No. NC3858392