Learn More
Research Products International Corp LB Broth Powder [Lennox L Broth], 1 Kilogram
Supplier: Research Products International Corp L240601000.0
For propagation and maintenance of E. Coli.Low Salt Formula.Molecular Biology Grade. Dissolve 20 grams in 1 liter distilled water. Then adjust pH to 7.2. Heat and frequently agitate to completely dissolve. Sterilize by autoclaving for 15 minutes. Ingredients: Tryptone: 10 g/L Yeast Extract: 5 g/L Sodium Chloride: 5 g/L
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.