Learn More
Aniara Diagnostica LLC LENS CULINARIS LECTIN 10 MG
Supplier:  Aniara Diagnostica LLC A05010410
Encompass
Medicago Lens culinaris lectin 10 mg is isolated from lentil seeds by affinity chromatography. Supplied without preservatives as a white lyophilized powder from 1 mM CaCl2, 1 mM MnCl2 and 1 mM MgCl2. Store at -20C.
Catalog No. 50-100-8769
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.