Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Lens Holder
SCI_ED

Catalog No. S24372 Shop All Eisco Products
Change view
Click to view available options
To hold lenses or mirros of 50mm dia.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S24372 To hold lenses or mirros of 50mm dia.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S24372 Supplier Eisco™ Supplier No. PH0550F
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

To hold lenses or mirrors of 50mm dia.

  • Strip metal spring clip with locating V in either side arm
  • Mounted on stand with groove to accommodate meter rule

Specifications

Product Type Lens Holder
For Use With (Equipment) Lenses or mirrors of 50 mm dia.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.