Learn More
Clontech Labs 3P Lenti-X™ Expression System (EF1alpha Version)
Supplier: Clontech Labs 3P 631253
The Lenti-X Expression System (EF1α Version) enables you to produce exceptionally high titers of recombinant lentivirus for expression of any cDNA in any cell type susceptible to lentiviral transduction. The System contains an HIV-1-based, lentiviral expression vector, pLVX-EF1α-IRES-Puro, designed to constitutively express a protein of interest and puromycin resistance (Puror). Constitutive expression of these proteins is driven by the human elongation factor 1 alpha (EF1α) promoter, which allows protein expression in mammalian cells in which the CMV promoter is silenced. The System also includes Lenti-X Packaging Single Shots (VSV-G) and Lenti-X GoStix.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.