Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, human, non-targeting, with GFP

Catalog No. p-7182874
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A32063 100 μL
A32831 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
For Research Use Only. Not for use in diagnostic procedures.
Catalog No. A32063 Supplier Invitrogen™ Supplier No. A32063
Only null left
Add to Cart
Edge
Add to Cart
For Research Use Only. Not for use in diagnostic procedures.

Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.

Negative control gRNAs are essential in screening applications for setting thresholds for hit determination.

The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

The Invitrogen™ LentiArray™ CRISPR Negative Control Lentivirus, Human Scrambled, contains a gRNA sequence with no sequence homology to any region of the human genome. This construct also expresses emGFP to provide a visual readout of successful transduction that can be used in determining Multiplicity of Infection (MOI).

Recommended Storage

Store at -68 to -85°C

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 1 x 100 μL. Store (at -68°C to -85°C.)
Delivery Type Lentiviral
Format Tube
Control Type Negative Control
RNAi Type gRNA
Species Human
For Use With (Application) Genome Editing
Product Line LentiArray
Product Type gRNA
Promoter U6
Quantity 100 μL
Shipping Condition Dry Ice
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.