Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LentiArray™ Human Ubiquitin CRISPR Library

Catalog No. A42270
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Library
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A42270 1 Library
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A42270 Supplier Invitrogen™ Supplier No. A42270
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The award-winning LentiArray Human Ubiquitin Library targets 943 genes with up to four gRNA per gene target pooled in a single well.

The award-winning LentiArray Human Ubiquitin Library targets 943 genes with up to four gRNA per gene target pooled in a single well. Libraries are delivered as 100 μL of ready-to-use lentiviral particles per gene target with an average titer of 1x108 TU/mL (functional titer determined by antibiotic resistance).

The ubiquitin system is integral to maintaining cellular homeostasis by regulating protein turnover. Dysregulation of the ubiquitin system has been linked to multiple diseases, including cancer, neurodegenerative, musculoskeletal, cardiovascular, and metabolic diseases, as well as being linked to viral infection. The gene targets within this library were selected using the most up-to-date genome databases, including the NCBI RefSeq database, and cross-referenced to the Gene Ontology Consortium (GO) database and/or the HUGO Gene Nomenclature Committee (HGNC).

  • 943 genes with up to four gRNA per gene target (pooled in a single well) for a total of 3,722 gRNAs
  • 100 μL of ready-to-use lentiviral particles per gene target
  • Average titer of 1x108 TU/mL. (functional titer determined by antibiotic resistance)

Built to bring you success

LentiArray CRISPR libraries are constructed using gRNA designs produced using Thermo Fisher Scientific's proprietary CRISPR gRNA design algorithm. This algorithm incorporates the latest gRNA design research, as well as our extensive in-house experience, to produce the highest quality designs. The gRNA designs included in the LentiArray CRISPR libraries are selected for maximal editing efficiency and specificity and are designed to knock out all known isoforms of the target gene. For each gene target, up to four high-quality gRNAs are included to help ensure highly efficient knockout of the target gene in a wide array of cell types.

Design and execute your experiments without limitations

LentiArray CRISPR libraries are designed and constructed to give you complete control over your experimental design. They utilize a two-vector design, expressing the Cas9 nuclease and the gRNA off of separate lentiviral constructs, enabling you to dictate when and how the genome editing tools are delivered to your cells.

The LentiArray Cas9 lentivirus construct expresses a human, codon-optimized Cas9 nuclease with a blasticidin-resistance gene under the control of the EF-1α (EFS) promoter. You can choose to co-infect your cells with both the LentiArray gRNA lentiviruses and the Cas9 lentivirus or just the LentiArray Cas9 lentivirus to establish a stable Cas9 expressing cell line. With LentiArray CRISPR libraries you have the freedom to design the experimental approach that best suits your model system and your screening goals.

Furthermore, to minimize restrictions to your experimental design, the gRNA lentiviruses that make up the LentiArray CRISPR libraries do not contain fluorescent markers. This allows reporters of any wavelength to be used with the libraries and expands your ability to perform multiplexed experiments. For challenging cell lines, the LentiArray gRNA lentiviruses contain a puromycin resistance gene that allows enrichment of the population of edited cells and drives stronger phenotypes.

TRUSTED_SUSTAINABILITY

Specifications

Product Line LentiArray
Quantity 1 Library
Content And Storage 1 library set, 100 μL/target.
Store at -68 to -85°C.
Delivery Type Lentiviral
Product Type Human Ubiquitin CRISPR Library
Promoter U6
Shipping Condition Dry Ice
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.