Learn More
Selleck Chemical LLC Lentinan 25mg 37339-90-5 A823605, Bromoduline
SDP
Supplier:  Selleck Chemical LLC S508325MG
Encompass_Preferred
Lentinan (A823605, Bromoduline), isolated and purified from a hot water extract of Lentinula edodes fruit bodies, is a fungal polysaccharide immunomodulator with anti-tumor effects. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8335
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.