Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mirus Bio™ Lentivirus Packaging Mix Powered by MISSION™ Genomics

Catalog No. p-8504044 Shop All Mirus Bio LLC Products
Change view
Click to view available options
No. of Reactions:
5
34
Volume (Metric) Sample:
0.25 mL
1.7 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions Volume (Metric) Sample
MIR6640 34 1.7 mL
MIR6630 5 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MIR6640 Supplier Mirus Bio™ Supplier No. MIR 6640
Only null left
Add to Cart
Add to Cart

Mirus Bio Lentivirus Packaging Mix Powered by MISSION™ Genomics is an optimized formulation of lentivirus vectors that enables the generation of high functional lentivirus in HEK 293 cells.

  • Accelerate viral production by skipping tedious but critical multivariable plasmid optimization.
  • Produce high titer lentivirus with the TransIT™-Lenti Transfection Reagent.
  • Compatible with second or third generation transfer vector encoding gene of interest (GOI).
  • Choose between small or large scale lentivirus production with our conveniently sized 5 or 34 reaction sizes for 10 cm dish.
  • Optimized formulation of two plasmids expressing the structural and regulatory (gag, pol, and rev) and viral envelope (VSV-G) proteins required for producing high titer lentivirus with broad viral tropism.

Specifications

Content And Storage -20°C
Cell Type 293-derived Cells
Format Kit
Sample Type Plasmid DNA
Promoter CMV
Transfection Technique Chemical Transfection
Concentration Proprietary
Vector pMISSION
Formulation Proprietary
For Use With (Application) Lentivirus production
No. of Reactions 34
Product Type Lentivirus packaging vectors
Quantity 1 Ea.
Sterility Sterile
Volume (Metric) Sample 1.7 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.