Learn More
Genscript Corporation Lentivirus Titer p24 ELISA Kit
Supplier:  Genscript Corporation L00974
Encompass_Preferred
Lentivirus Titer p24 ELISA Kit Pro consists of two parts including Lentivirus Free p24 Removal Reagent (patented technology) and Lentivirus Titer p24 ELISA Kit Before detecting the concentration of p24 protein free p24 in lentiviral supernatant can be specifically removed by Lentivirus Free p24 Removal Reagent
Catalog No. 50-000-58398
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.