Learn More
United Scientific™ Lenz's Law Apparatus
SDPSCI_ED
Catalog No. S000866 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Dimensions (L x W x H):
3 x 8 x 6 in.
Catalog No. Dimensions (L x W x H)
S000866 3 x 8 x 6 in.
1 options
Prop 65
Catalog No. S000866 Supplier United Scientific™ Supplier No. LENZLAW
Add to Cart
Add to Cart
Prop 65
Demonstrate the principles of magnetic induction with the Lenz’s Law Apparatus.
The central pointed support balances an aluminum arm that has two aluminum loops on the ends. One loop is solid while the other has a slit. When a magnet is brought near a metal loop it induces an opposing magnetic field. When the loop is incomplete, an induced current cannot be created. This causes the complete loop to experience motion, while the incomplete loop does not move. Apparatus measures 3 x 8 x 6 in. (7.6 x 20 x 15.2 cm).
  • Pointed support evenly balances the arm
  • Aluminum design is ferromagnetic but can have an induced magnetic field
  • The device is sensitive enough to work with a variety of magnets (not included), but neodymium magnets yield the best results.
  • Includes activity guide
Specifications
Grade 5+
Product Type Lenz Law Kit
Dimensions (L x W x H) 3 x 8 x 6 in.