Learn More
MyBioSource LEPR elisa kit
SDP
Supplier:  MyBioSource MBS202335896STRIPWELLS
Encompass_Preferred
The MyBioSource Human LEPR ELISA Kit quantifies human Leptin Receptor in various biological fluids using a 96-well plate coated with a specific antibody. Samples and standards are added, followed by a biotinylated detection antibody, HRP-conjugated streptavidin, and a TMB substrate that yields a color change measured at 450 nm.
Catalog No. NC2601730
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.