Learn More
Crystal Chem Inc Leptin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80968
Encompass_Preferred
The Leptin ELISA is a highly sensitive assay that is used to quantify levels of human leptin in serum. Highlights: Kit uses sample size of only 20 µL. High sensitivity of 0.42 ng/mL. Wide dynamic range of 1- 100 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of ELISA kits for human markers. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7871
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.