Learn More
CPC Scientific H-Asn-Val-Ile-Gln-Ile-Ser-Asn-Asp-Leu-Glu-Asn-Leu-Arg-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LEPT001B
Encompass
ONE-LETTER SEQUENCE: NVIQISNDLENLRMOLECULAR FORMULA: C64H110N20O23MOLECULAR WEIGHT:1527.70STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [200436-43-7], SYNONYMS: Obese Gene Peptide (93-105) (human)RESEARCH AREA: ObesityREFERENCES: Y. Zhang et al., Nature, 372, 425 (1994); M.A. Pelleymounter et al., Science 269, 541 (1995)
Catalog No. 50-210-9632
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.