Learn More
CPC Scientific H-Val-Pro-Ile-Gln-Lys-Val-Gln-Asp-Asp-Thr-Lys-Thr-Leu-Ile-Lys-Thr-Ile-Val-Thr-Arg-Ile-Asn-Asp-Ile-Ser-His-Thr-Gln-Ser-Val-Ser-Ser-Lys-Gln-Lys-OH (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific LEPT003B
Encompass
Leptin administration produces fragments of rat inhibition of food intake. ONE-LETTER SEQUENCE: VPIQKVQDDTKTLIKTIVTRINDISHTQSVSSKQKMOLECULAR FORMULA: C171H298N50O56MOLECULAR WEIGHT:3950.55STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [183598-56-3], SYNONYMS: Obese Gene Peptide (22-56) (human)RESEARCH AREA: ObesityREFERENCES: Y. Zhang et al., Nature, 372, 425 (1994); M.A. Pelleymounter et al., Science 269, 541 (1995)
Catalog No. 50-210-9636