Learn More
Description
Specifications
Specifications
| Assay Range | 7.81 pg/mL |
| Assay Sensitivity | 7.81-500 pg/mL |
| Assay | Sandwich ELISA |
| Conjugate | HRP |
| Product Type | Antibody Pairs |
| Specificity | This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human Leptin/OB. |
| Target Species | Human |
| For Use With (Application) | Sandwich ELISA |
| Gene Alias | FLJ94114, leptin, leptin (murine obesity homolog), leptin (obesity homolog, mouse), Obese protein, obese, mouse, homolog of, Obesity factor, OBOBS |
| Gene ID (Entrez) | 3952 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
