Learn More
Invitrogen™ Leptin Polyclonal Antibody

Description
PA1-9571 detects Leptin in mouse and rat samples. PA1-9571 has been successfully used in Western blot and ELISA procedures. By Western blot, PA1-9571 detects a 16 kDa protein representing Leptin. PA1-9571 immunogen is a purified recombinant rat Leptin protein.
Specifications
Specifications
| Antigen | Leptin |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | Lep |
| Gene Accession No. | P41160, P50596 |
| Gene Alias | CD295 antigen; H-Leptin; HuB219; Lep; LEPD; LEP-R; Leptin; leptin (murine obesity homolog); leptin (obesity homolog, mouse); leptin precursor; Method: conceptual translation supplied by author.; OB; obese; obese protein; obese, mouse, homolog of; obesity; obesity factor; obesity protein; OB-R; OBS; truncated leptin |
| Gene Symbols | Lep |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.