Learn More
Valutek™ Letter-Size Cleanroom Paper
Catalog No. 191502132
Change view
Click to view available options
Color:
Blue
Green
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
19-150-2132 White
19-150-2131 Blue
19-150-2133 Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 19-150-2132 Supplier Valutek™ Supplier No. VTCRPS23W8511
Only null left
Add to Cart
Add to Cart
23lb letter size cleanroom paper is specially formulated with cellulose fiber, surrounded and bonded by a polymer impregnated substrate. This special formulation minimizes particulate and ink smearing and provides the paper with high tear and tensile strength
  • 23lb letter size
  • 250 sheets/ream, 10 reams/case, 2500 sheets/case.
  • 8.5 x 11 in. , (21.5 x 28 cm)
  • Ultra-low levels of particle and extractable counts
  • This product is commonly used for note-taking, laser printing, and photocopies. It is also used as a form and log sheet in a cleanroom Class 1 -10 (ISO 3-4) critical environment
  • It is compatible with most inks without smearing while ensuring low particle and fiber generation and low ionic contamination. 
  • All Valutek™ cleanroom papers are tested and manufactured in ISO compliant facilities under Valutek™ inspection and strict process control to ensure Valutek™ quality standard and product specifications
Specifications
Material Cellulose
Color White
Cover Material Polymer impregnated substrate
Size Letter