Learn More
Thermo Scientific Chemicals [Leu8] Renin Substrate Tetradecapeptide, Rat
For cellular and molecular biology applications
$360.00
Specifications
Color | White to off-white |
---|---|
Form | Solid |
Molecular Weight (g/mol) | 1789.06 |
Storage Requirements | -30° to -10°C |
Catalog Number | Mfr. No. | Substrate Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Substrate Quantity | Price | Quantity & Availability | |||||
AAJ66131LB0
|
Thermo Scientific Chemicals
J66131LB0 |
5mg |
Each of 1 for $360.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White to off-white | |
Solid | |
1789.06 | |
-30° to -10°C |
Safety and Handling
Recommended Storage : Store at -20°C
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.