Learn More
Novus Biologicals™ Leucine Aminopeptidase (LAP) Activity Assay Kit (Fluorometric)
Shop All Novus Products

Description
In this assay, LAPs hydrolyze leucine from the fluorescent probe and the amount of fluorescent probe detected is used to determine the total activity of the LAP enzymes. Detection method: Fluorescence (Ex/Em = 368/460 nm). The assay is simple to perform, high-throughput adaptable and can detect less than 0.1 mU of LAP activity.
Kit Contents:
- AMC-Standard (1 mM)
- LAP Assay Buffer
- LAP Positive Control
- LAP Substrate
Specifications
Specifications
| Content And Storage | Store at -20°C in the dark. Avoid freeze-thaw cycles. |
| Quantity | 100 Assays |
| Target | Leucine Aminopeptidase (LAP) Activity |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.