Learn More
CPC Scientific H-Asp-Pro-Ala-Phe-Asn-Ser-Trp-Gly-NH2 (trifluoroacetate salt) 5MG
SDP
Supplier:  CPC Scientific LEUC001B
Encompass
Leucokinin I, DPAFNSWG-amide, stimulated rapid fluid secretion in the Malpighian tubules of Manduca sexta, the mosquito Aedes egypti, the cricket Acheta domestica and in adult Drosophila melanogaster.ONE-LETTER SEQUENCE: DPAFNSWGMOLECULAR FORMULA: C41H53N11O12MOLECULAR WEIGHT:891.94STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [104600-89-7], RESEARCH AREA: MiscellaneousREFERENCES: N.J.V.Skaer et al., J. Exp. Biol., 205, 1869 (2002)
Catalog No. 50-210-9642
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.