Learn More
Greiner Bio-One Leucosep Polypropylene Centrifugation Tubes
GREENER_CHOICE
Catalog No. 07000448 Shop All Greiner Bio-One Products
Change view
Click to view available options
Capacity (Metric):
12 mL
50 mL
Sterility:
Aseptically Manufactured
Sterile
Bottom Shape:
Conical
Round
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Sterility Bottom Shape
07-000-448 12 mL Aseptically Manufactured Round
07-000-449 12 mL Sterile Round
07-000-499 50 mL Aseptically Manufactured Conical
07-000-500 50 mL Sterile Conical
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 07-000-448 Supplier Greiner Bio-One Supplier No. 163288
Add to Cart
Add to Cart
Request a Quote
For efficient separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow.
  • Enrichment directly from whole blood
  • Simplified filling through porous barrier
  • Rapid separation in 15 minutes at room temperature
  • Removal of erythrocytes and granulocytes
  • No recontamination with erythrocytes
  • Service Plan
    No
    TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 12 mL
Material Polypropylene
For Use With Separation of lymphocytes and mononuclear cells from peripheral blood and bone marrow
Includes Porous barrier
Sterility Aseptically Manufactured
Bottom Shape Round
Product Type Centrifuge Tube (Pre-filled)