Learn More
Description
Leucovorin Calcium, USP is an odorless, hygroscopic powder that ranges from yellowish-white to yellow in color.
Premium Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered drug manufacturer. Qualified manufacturers adhere to FDA registration and comply with general facility and equipment-specific GMP standards (21 CFR Parts 210 and 211). Compliance with stability and testing may vary. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.
Alternative name(s): Calcium folinate, Folinic acid calcium salt
Specifications
Specifications
| Chemical Name or Material | Leucovorin Calcium, USP |
| Packaging | Bottle, Amber Glass |
| CAS | 1492-18-8 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 511.5 |
| Quantity | 1 g |
| Percent Purity | 95 to 105 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
