Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medisca Leucovorin Calcium, USP
SDP

Catalog No. 14186432 Shop All Medisca Products
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
25 g
500 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-186-432 500 g
14-185-613 1 g
14-185-614 100 mg
14-185-421 5 g
14-185-445 500 mg
14-185-377 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 14-186-432 Supplier Medisca Supplier No. 017808
Add to Cart
Add to Cart

Leucovorin Calcium, USP is an odorless, hygroscopic powder that ranges from yellowish-white to yellow in color.

Leucovorin Calcium, USP is an odorless, hygroscopic powder that ranges from yellowish-white to yellow in color.

Premium Supplement Grade refers to a nutritional supplement that is manufactured by an FDA-registered drug manufacturer. Qualified manufacturers adhere to FDA registration and comply with general facility and equipment-specific GMP standards (21 CFR Parts 210 and 211). Compliance with stability and testing may vary. The customer is responsible for ensuring suitability of the product for the intended use and acknowledges that further testing may be required.

Alternative name(s): Calcium folinate, Folinic acid calcium salt

Specifications

Chemical Name or Material Leucovorin Calcium, USP
Packaging Jar 1500ml, White, HDPE, Round, 89-400, TRIVENI-DB, CT
CAS 1492-18-8
Type Drug Standard
Molecular Weight (g/mol) 511.5
Quantity 500 g
Percent Purity 95 to 105 %
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.