Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Leukocyte Aggregation Inhibitor

Catalog No. 89339173 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-173 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-173 Supplier Abnova™ Supplier No. DU0008
Add to Cart
Add to Cart

Leukocyte Aggregation Inhibitor suppresses leukocyte aggregation

Specifications

For Use With (Application) IWA
Content And Storage 4°C
Quantity 150 μL
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.