Learn More
Enzo Life Sciences Leupeptin (synthetic) (5 mg) CAS: 103476-89-7
Supplier: Enzo Life Sciences ALX260009M005
Cathepsin B inhibitor. Competitive and reversible inhibitor of serine and cysteine proteinases like calpain, porcine kallikrein, trypsin, plasmin, papain and cathepsin B. Does not inhibit chymotrypsin and thrombin. Also known to inhibit activation-induced programmed cell death and restoration of defective immune responses of HIV+ donors. Alternative Name: Acetyl-L-leucyl-L-leucyl-L-argininal . hemisulfate. Formula: C20H38N6O4. 0.5 H2SO4. MW: 426.6. 49.0. CAS: 103476-89-7. MI: 14: 5456. Purity: ≥97% (HPLC). Appearance: White to off-white powder. Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: -20°C. Handling: Protect from light and moisture.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.