Learn More
Description
When adjusted for molarity, all three salt forms are equally effective; however, the hydrochloride salt is usually less invasive in biological settings.
Inhibits trypsin-like proteases and some cysteine proteases including: endoproteinase Lys-C, kallikrein, papain, thrombin, cathepsin B, cathepsins H and L, trypsin, calpain, trypsin8 and plasmin.
Specifications
Specifications
| CAS | 103476-89-7 |
| Content And Storage | Store at −20°C, desiccate, protect from light. |
| Format | White Powder |
| Molecular Weight (g/mol) | 427.57 |
| Quantity | 25mg |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.