Learn More
Promega LEV Plungers
Catalog No. PRAS6101 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
50/Pk.
Catalog No. Quantity
PRAS6101 50/Pk.
1 options
Catalog No. PRAS6101 Supplier Promega Supplier No. AS6101
LEV Plungers for Maxwell™ 16 Instruments in low elution volume configuration.
  • Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Standard Elution Volume (SEV) is 200-400μL
  • Low Elution Volume (LEV) is 30-100μL
  • Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)
Specifications
Content And Storage 15°C to 30°C
Isolation Technology Magnetic Bead
Quantity 50/Pk.