Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

High-Tech Conversions Bee-Safe™ Isolation Gowns

Catalog No. 17444082
Change view
Click to view available options
Size:
Large
X-Large
Color:
Blue
Yellow
Style:
Level 1
Level 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Color Style
17-444-082 X-Large Yellow Level 1
17-444-081 Large Yellow Level 1
17-444-084 X-Large Blue Level 2
17-444-083 Large Blue Level 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 17-444-082 Supplier High-Tech Conversions Supplier No. GAHGISO1YX

Spunbond polypropylene isolation gowns are fluid resistant and ideal for a variety of healthcare settings. Choose from AAMI Level 1 or Level 2 styles.

  • TESTING: Level 1 Gown
    • Liquid Barrier: ANSI/AAMI PB70
    • Water Resistance (Impact Penetration Test): AATCC 42
    • Water Resistance (Hydrostatic Pressure Test): AATCC 127
  • TESTING: Level 2 Gown
    • Liquid Barrier: ANSI/AAMI PB70
    • Water Resistance (Impact Penetration Test): AATCC 42
    • Water Resistance (Hydrostatic Pressure Test): AATCC 127

Specifications

Gender Unisex
No. of Pockets None
Style Level 1
Product Type Isolation Gown
Size X-Large
For Use With (Application) Healthcare Settings
Material Laminated Spunbond Polypropylene
Color Yellow
Disposable Single-use
Sterility Non-sterile
Cuff Style Knit
Latex Free Yes
Model GAH-GISO1YX
Sleeve Style Long Sleeve
Certifications/Compliance ANSI/AAMI PB70, AATCC 42, AATCC 127
Autoclavable Not Autoclavable
No. per Pack 10/Bag, 5 Bags/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.