Description
- SMS non-woven material
- Knitting and elastic cuff for more protection
- Fluid resistant
- Color: white
- AAMI Level 2 standard
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | X-Large |
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Cuff Style | Knit |
| Sterility | Non-sterile |
| Weight (Metric) | 37 gsm |