Learn More
Apexbio Technology LLC Levomefolate calcium 151533-22-1 10mg
SDP
Supplier:  Apexbio Technology LLC A355210
Encompass_Preferred
Levomefolate calcium (CAS 151533-22-1) is the calcium salt form of L-5-methyltetrahydrofolate a metabolically active folate derivative Unlike folic acid it does not require enzymatic reduction and methylation in vivo allowing direct participation in folate-dependent one-carbon transfer reactions involved in homocysteine remethylation and DNA synthesis Due to its enhanced bioavailability and immediate coenzymatic function levomefolate calcium is frequently applied in biomedical research to investigate folate metabolism methylation pathways and the potential impact of folate status in neuropsychiatric disorders cardiovascular health and cellular proliferation models
Catalog No. 50-000-94801
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.