Learn More
Medchemexpress LLC HY-B1693 10mg Medchemexpress, Levomepromazine CAS:60-99-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB169310MG
Encompass_Preferred
Medchemexpress, HY-B1693 10mg Levomepromazine CAS:60-99-1 Methotrimeprazine (Levomepromazine) is an orally available neuroleptic agent, which is commonly used to relieve nausea and vomiting in palliative care settings. Levomepromazine has antagonist actions at multiple neurotransmitter receptor sites, including dopaminergic, cholinergic, serotonin and histamine receptors [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.