Learn More
Selleck Chemical LLC Levosimendan S2446-1g
SDP
Supplier:  Selleck Chemical LLC S24461G
Encompass_Preferred
Levosimendan(OR1259 OR1855 Simsndan) is a calcium sensitizer acting through calcium-dependent binding to cardiac troponin C (cTnC) provides treatment for heart failure
Catalog No. 50-311-2390
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.