Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Lewis A/B Chimeric Recombinant Rabbit Monoclonal Antibody (HEA164)
GREENER_CHOICE

Catalog No. PIMA548240
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548240 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548240 Supplier Invitrogen™ Supplier No. MA548240
Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody is specific for Lewis a,b carbohydrate antigens.

Lewis A and Lewis B are fucose-containing carbohydrates that are expressed on the surface of red blood cells and other tissues in the body. These carbohydrates are part of the larger family of molecules known as Lewis antigens, which play a role in a variety of biological processes, including immune recognition, cell adhesion, and inflammation. The expression of Lewis A and B on red blood cells is determined by the FUT3 gene, which encodes for the fucosyltransferase enzyme that adds the fucose sugar to precursor molecules. In general, individuals with the FUT3 gene mutation that leads to the expression of Lewis A antigen are unable to produce the Lewis B antigen, and vice versa. However, some individuals may be Lewis negative, meaning that they do not express either antigen due to mutations in the FUT2 gene. The presence or absence of Lewis antigens can also have implications for disease susceptibility and treatment, particularly in the context of cancer and other inflammatory conditions, as they can modulate immune responses and promote tumor growth and metastasis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Lewis A/B Chimeric
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HEA164
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene EPCAM
Gene Accession No. P16422
Gene Alias Lewis A; Lewis B
Host Species Rabbit
Immunogen This antibody was raised by immunising mice with human carcinoma cells.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4072
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.