Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Leyden Jar
SDPSCI_ED

Catalog No. S139486 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139486 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139486 Supplier United Scientific™ Supplier No. LYJ001
Add to Cart
Add to Cart
This item is not returnable. View return policy

Demonstration of static electricity.

Learn about static electricity and the function of a dielectric with this device.

  • Two aluminum jars (one with an attached aluminum electrode) and one plastic jar are used to collect the charge
  • Assembled device is 6 in. tall and 3-1/4 in. in diameter
  • Includes activity guide

Specifications

Grade 5 to 12
Format Kit
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.