Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LFC Sheep anti-Mouse, Polyclonal, Exalpha
SDP

Catalog No. 502101885
Encompass
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-210-1885 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-210-1885 Supplier Vector Laboratories Supplier No. X1089P
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

LFC Polyclonal antibody specifically detects LFC in Mouse samples. It is validated for Immunofluorescence, Immunoprecipitation, Western Blot

Specifications

Antigen LFC
Applications Immunofluorescence, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.08% sodium azide; pH
Gene ARHGEF2
Gene Accession No. Q60875
Gene Alias AA408978; Arhgef2; CG10188; CG10188 gene product from transcript CG10188-RB; CG10188-PA; CG10188-PB; CG10188-PC; DKFZp547L106; DKFZp547P1516; Dmel\CG10188; Dmel_CG10188; FLJ45440; GEF; GEFH1; GEF-H1; Guanine nucleotide exchange factor H1; Kiaa0651; Lbcl1; LBC'S first cousin; Lfc; LFP40; Lymphoid blast crisis-like 1; microtubule-regulated Rho-GEF; mKIAA0651; Oncogene LFC; OTTHUMP00000018855; P40; Proliferating cell nucleolar antigen p40; rho guanine nucleotide exchange factor 2; rho guanine nucleotide exchange factor 2-like protein; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rho/Rac guanine nucleotide exchange factor 2; rhobin; uncharacterized protein
Host Species Sheep
Immunogen Antibody developed using protein corresponding to amino acids 2 to 447 of the Lfc protein fused to GST.
Purification Method Ammonium sulfate precipitation
Quantity 250 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16800
Target Species Mouse
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.