Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
LG 101506, Tocris Bioscience™
Description
Selective RXR modulator (Ki values are 3, 9 and 11 nM for RXRα, RXRβ and RXRγ respectively). Displays poor binding affinity for RAR isoforms (Ki values are 2746, 3516 and >10,000 nM for RARα, RARβ and RARγ respectively). Binding to RXR results in selective activation of RXR:PPARγ, RXR:PPARα and RXR:PPARδ heterodimers.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 420.53 |
| Percent Purity | >98% |
| Chemical Name or Material | LG 101506 |
Safety and Handling
| Recommended Storage | Store at +4°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?