Learn More
JDM SCIENTIFIC LLC Large Capacity Digital Tube Rotator
Supplier:  JDM SCIENTIFIC LLC TR05UA
Encompass_Preferred
The title angle is adjustable for accommodating different applicationsVariable sizes of Tube Holders and Carousels are available to accommodate various sizes of TubesAll units are compatible with a cold room and incubator environmentTR-04UA and TR-05UA models have speed adjustment, continuous and timer mode with automatic stop
Catalog No. NC2385824
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.