Learn More
Description
- Human LGALS12 full-length ORF ( AAH28222, 1 a.a. - 336 a.a.) recombinant protein with GST-tag at N-terminal
- Theoretical molecular weight: 62.7kDa
- Preparation: in vitro wheat germ expression system
- Purification: glutathione sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced glutathione, pH: 8.0 in elution buffer
Sequence: MSQPSGGRAPGTRIYSWSCPTVMSPGEKLDPIPDSFILQPPVFHPVVPYVTTIFGGLHAGKMVMLQGVVPLDAHRFQVDFQCGCSLCPRPDIAFHFNPRFHTTKPHVICNTLHGGRWQREARWPHLALRRGSSFLILFLFGNEEVKVSVNGQHFLHFRYRLPLSHVDTLGIFGDILVEAVGF LNINPFVEGSREYPAGHPFLLMSPRLEVPCSHALPQGLSPGQVIIVRGLVLQEPKHFTVSLRDQAAHAPVTLRASFADRTLAWISRWGQKKLISAPFLFYPQRFFEVLLLFQEGGLKLALNGQGLGATSMNQQALEQLRELRISGSVQLYCVHS
Best use within three months from the date of receipt.
ELISA, western blotting (recombinant protein), antibody production, protein array
Specifications
Specifications
| For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
| Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| Molecular Weight (g/mol) | 62.7 |
| Name | Human LGALS12 Full-length ORF Recombinant Protein with GST-tag at N-terminal |
| pH Range | 8 |
| Preparation Method | In vitro wheat germ expression system |
| Purification Method | Glutathione Sepharose 4 Fast Flow |
| Quality Control Testing | 12.5% SDS-PAGE stained with Coomassie Blue |
| Quantity | 25 μg |
| Source | Wheat Germ (in vitro) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.