Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ LGALS3 (Mouse) ELISA Kit

Catalog No. 89100427 Shop All Abnova Corporation Products
Change view
Click to view available options
Sample Type:
CCS, Plasma (EDTA, Heparin), Serum
Serum, Plasma, CCS, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Type
89-100-427 Serum, Plasma, CCS, Urine
89-101-048 CCS, Plasma (EDTA, Heparin), Serum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-100-427 Supplier Abnova™ Supplier No. KA2147

Includes

  • Galectin-3 Microplate (Item A): 96 wells coated with anti-mouse Galectin-3
  • Wash Buffer Concentrate (20x) (Item B)
  • Standards (Item C): recombinant mouse Galectin-3
  • Assay Diluent A (Item D)
  • Assay Diluent B (Item E)
  • Detection Antibody Galectin-3 (Item F): 2 vial of biotinylated antimouse Galectin-3 (each vial is enough to assay half microplate
  • HRP-Streptavidin Concentrate (Item G): 8μL 20,000x concentrated HRP-conjugated streptavidin
  • TMB One-Step Substrate Reagent (Item H): 12mL of 3,3',5,5'-tetramethylbenzidine (TMB) in buffer solution
  • Stop Solution (Item I): 2M sulfuric acid

Use to measure quantitatively mouse Lgals3. Sandwich enzyme immunoassay.

This assay employs an antibody specific for mouse Galectin-3 coated on a 96-well plate

Specifications

Conjugate HRP
Cross Reactivity Mouse
Description Lgals3 (Mouse) ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of mouse Lgals3.
Detection Limit 6pg/mL
Detection Method Colorimetric
Includes Antibody coated microwells (96 wells), Wash Buffer Cincentrate (20x) (ItemB): 25mL, Standard (Item C): 2 vials, Assay Diluent A (Item D): 30mL, Assay Diluent B (Item E): 15mL, Detection Antibody MSP (Item F): 2 vials, HRP-Streptavidian concentrrate (ItemG): 8μL, TMB One-Step Substrate Reagent (ItemH): 13mL, Stop solution (Item I): 8mL
Regulatory Status For research use only (RUO)
Sample Type Serum, Plasma, CCS, Urine
Sample Volume 100 μL
Sensitivity 6 pg/mL
Packaging 96-well
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type Lgals3 (Mouse) ELISA Kit
No. of Assays 96
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.