Learn More
Invitrogen™ LGALS3BP Monoclonal Antibody (D5)

Description
LGALS3BP Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
Specifications
| Antigen | LGALS3BP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | D5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 |
| Gene | LGALS3BP |
| Gene Accession No. | O70513, Q08380 |
| Gene Alias | 90K; basement membrane autoantigen p105; BTBD17B; CyCAP; Cyp-C-associated protein; galectin 3 binding protein; galectin-3-binding protein; gp90; L3 antigen; Lectin; lectin galactoside-binding soluble 3 binding protein; lectin galactoside-binding soluble 3-binding protein; lectin, galactoside binding soluble 3 binding protein; lectin, galactoside-binding, soluble, 3 binding protein; Lgals3bp; M2BP; mac-2 BP; mac-2-binding protein; MAC2BP; MAC-2BP; MAC-2-BP; mama; peptidylprolyl isomerase C-associated protein; Ppicap; Protein 90K; Protein MAMA; serum protein 90K; similar to Galectin-3 binding protein precursor (Lectin galactoside-binding soluble 3 binding protein) (Mac-2 binding protein) (Mac-2 BP) (MAC2BP) (Tumor-associated antigen 90K); TANGO10B; transport and golgi organization 10 homolog B; tumor-associated antigen 90K |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.