Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Corporation LgBiT Expression Vector

Catalog No. PRN2681 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRN2681 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRN2681 Supplier Promega Corporation Supplier No. N2681
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Perform live-cell, kinetic analysis of HiBiT-tagged proteins using Express Intracellular LgBiT Protein for HiBiT Analysis, without cell lysis. Compatible with exogenously or endogenously expressed HiBiT fusion proteins, and ideal for live-cell NanoBRET™ applications.

  • Simple and effective RNA/DNA extraction from biological samples.
  • High yield and purity of nucleic acids.
  • Suitable for various downstream applications including PCR.
  • Consistent performance with minimal variability.
  • Compatible with multiple sample types.
  • Streamlined protocol for quick processing.
  • Eliminates the need for hazardous chemicals.
  • Cost-effective solution for nucleic acid extraction.
  • Reliable and reproducible results.
  • Easy integration into existing workflows.

Specifications

Detection Method Nano-Glo™ Live Cell Substrates
Product Type Expression Vector
Species Oplophorus gracilirostris
Protein Tag HiBit
Concentration 1 μg, μL
Label or Dye NanoLuc™ luciferase
Quantity 1 Ea.
Vector LgBit
Color Translucent
For Use With (Application) Real time monitoring of protein interactions, protein dynamics studies, reporter gene assays, drug screening
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.