Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGMN Recombinant Rabbit Monoclonal Antibody (K01_1T44)
GREENER_CHOICE

Catalog No. MA563404
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA563404 100 μL
MA564863 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA563404 Supplier Invitrogen™ Supplier No. MA563404
  • $544.00
    $408.00 / Each of 1

    Save $136.00

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: VYATTAANPR ESSYACYYDE KRSTYLGDWY SVNWMEDSDV EDLTKETLHK QYHLVKSHTN TSHVMQYGNK TISTMKVMQF QGMKRKASSP VPLPPVTHLD LTPSPDVP Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%, Pig - 86%, Cynomolgus monkey - 97%.

This gene encodes a cysteine protease that has a strict specificity for hydrolysis of asparaginyl bonds. This enzyme may be involved in the processing of bacterial peptides and endogenous proteins for MHC class II presentation in the lysosomal/endosomal systems. Enzyme activation is triggered by acidic pH and appears to be autocatalytic. Protein expression occurs after monocytes differentiate into dendritic cells. A fully mature, active enzyme is produced following lipopolysaccharide expression in mature dendritic cells. Overexpression of this gene may be associated with the majority of solid tumor types. This gene has a pseudogene on chromosome 13. Several alternatively spliced transcript variants have been described, but the biological validity of only two has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGMN
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_1T44
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 150mM NaCl, 40% glycerol and 0.01% sodium azide; pH 7.4
Gene LGMN
Gene Accession No. Q99538
Gene Alias AEP; AI746452; Asparaginyl endopeptidase; AU022324; cb173; cb460; cysteine protease 1; legumain; Lgmn; LGMN1; preprolegumain; protease, cysteine 1; protease, cysteine, 1; protease, cysteine, 1 (legumain); PRSC1; PRSC1EC 3.4.22.34; sb:cb173; wu:fa98g05; wu:fj92g08; zgc:76953
Gene Symbols LGMN
Host Species Rabbit
Immunogen Recombinant protein of human Legumain (aa 204-311).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5641
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.