Learn More
Invitrogen™ LGR4 Polyclonal Antibody

Description
PA1-20450 detects G Protein-Coupled Receptor GPR48 in human samples.
Specifications
Specifications
| Antigen | LGR4 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide |
| Gene | LGR4 |
| Gene Accession No. | Q9BXB1 |
| Gene Alias | 9130225G07; A330106J01Rik; A930009A08Rik; BNMD17; G protein-coupled receptor 48; G protein-coupled receptor GPR48; GPR48; G-protein coupled receptor 48; leucine rich repeat containing G protein-coupled receptor 4; leucine-rich repeat containing G protein-coupled receptor 4; leucine-rich repeat-containing G protein-coupled receptor 4; leucine-rich repeat-containing G-protein coupled receptor 4; Lgr4 |
| Gene Symbols | LGR4 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.