Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ LGR6 Recombinant Rabbit Monoclonal Antibody (HL2827)
GREENER_CHOICE

Catalog No. MA556779
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA556779 Supplier Invitrogen™ Supplier No. MA556779
  • $522.00
    $391.50 / Each of 1

    Save $130.50

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

LGR6 Recombinant Monoclonal Antibody for Western Blot, IHC (P)

LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LGR6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone HL2827
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Lgr6
Gene Accession No. Q9HBX8
Gene Alias A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631
Gene Symbols Lgr6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the Intracellular domain of human LGR6. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59352
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.