Learn More
Apexbio Technology LLC LGX818, 1mg. Cas: 1269440-17-6 MFCD: MFCD25976758. RAF inhibitor,potent and selective
SDP
Supplier:  Apexbio Technology LLC B11741
Encompass_Preferred
LGX818 is a potent and selective inhibitor of RAF kinase with EC50 value of 3nM in A375 cells [1].The V600E mutation of B-Raf in the kinase domain results in constitutive activation of B-Raf and promotes cancer development. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5209
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.