Learn More
Thermo Scientific™ LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
GSA_VA
Catalog No. 75006421
Change view
Click to view available options
Type:
Bucket Rotor
Double Spin Bucket
Microplate Adapter
Sealing Cap
Swinging Bucket Rotor
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
75-006-421 Sealing Cap
75-006-476 Swinging Bucket Rotor
75-006-477 Bucket Rotor
75-006-478 Double Spin Bucket
75-207-110 Microplate Adapter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 75-006-421 Supplier Thermo Scientific™ Supplier No. 75006421
Add to Cart
Add to Cart
Support a diverse array of sample processing applications without the need to modify existing protocols.
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
  • Capacity: 8 × 500mL (4L)
  • Max. Speed/RCF: 4400rpm/ 5,480 x g
Ordering Information:
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific Cryofuge 5500i series centrifuges
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Type Sealing Cap
For Use With (Equipment) Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Biocontainment Yes