Learn More
Thermo Scientific™ LH-4000W 8 x 500 mL Windshielded Swinging Bucket Rotor
Catalog No. 75207110 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Bucket Rotor
Double Spin Bucket
Microplate Adapter
Sealing Cap
Swinging Bucket Rotor
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
75-207-110 Microplate Adapter
75-006-421 Sealing Cap
75-006-476 Swinging Bucket Rotor
75-006-477 Bucket Rotor
75-006-478 Double Spin Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 75-207-110 Supplier Thermo Scientific™ Supplier No. 75006486
Add to Cart
Add to Cart
Support a diverse array of sample processing applications without the need to modify existing protocols.
Support a diverse array of sample processing applications without the need to modify existing protocols with the Thermo Scientific™ LH-4000W 8 x 500mL Windshield Swinging Bucket Rotor. Choose from round, DoubleSpin™ and Double blood bag buckets with a wide array of available adapters, providing capacity flexibility ranging from 1.5 to 250mL.
  • Capacity: 8 × 500mL (4L)
  • Max. Speed/RCF: 4400rpm/ 5,480 x g
Ordering Information:
Buckets, biocontainment lids and DoubleSpin buckets are sold separately; order rotor body (75006476) and components to complete assembly. Tube adapters and microplate adapter for DoubleSpin bucket are also available separately. Contact your Sales Representative for information.
Compatible with:
Thermo Scientific Cryofuge 5500i series centrifuges
TRUSTED_SUSTAINABILITY
Specifications
Type Microplate Adapter
For Use With (Equipment) Thermo Scientific Sorvall RC 4 Series, Cryofuge 5500i and Multifuge 4 Large Capacity Centrifuges
Biocontainment Yes