Learn More
Eppendorf 3P LI BATTERY MINI CR (P0625-21)
Supplier: Eppendorf 3P P06252100
The chart recorder will continuously monitor and record freezer temperature. It is front mounted to your freezer for easy access and can be fitted to all freezer models. • Includes chart recorder, RTD probe, keys, chart recorder paper, and chart recorder pens • Will continuously monitor and record freezer temperature for data documentation • Front mounted to your freezer for easy access and can be fitted to all freezer models • Battery powered: Will provide a minimum of eighteen months of operation • Selectable temperature range (0 to -50 oC and -50 to -100 oC) and rotation speed • User configurable built in high and low temperature alarms • Can be factory installed with your freezer order at no additional charge or retrofitted on location